Learn More
Hanna Instruments™ Bench Top pH Meters
Featuring a built-in 500 rpm magnetic stirrer, extended pH range, dual-level LCD with icons for stability and buffer recognition, and temperature display in either Celsius or Fahrenheit; this pH benchtop meter is ideal for educational purposes.
Brand: Hanna Instruments™ HI-207
This item is not returnable.
View return policy
Description
A low cost, space efficient bench top pH meter, with a built-in magnetic stirrer. The HI-208 also features automatic pH calibration, automatic temperature compensation and is supplied complete with pH electrode which incorporates the temperature sensor. Powered by mains electricity or the supplied 9V battery (run time up to 500hr).- Compact bench pH meter
- Built-in magnetic stirrer
- Mains or batteries operated
- Built-in sample beaker and electrode holder
- Dual level LCD display for pH and temperature
- pH electrode with temperature sensor included
- Automatic pH calibration and temperature compensation
Specifications
Bench Top pH Meter | |
±0.02 | |
Automatic | |
0.01 | |
-5.0°C to +105.0°C | |
LCD | |
HI207 | |
-2 to 16 pH | |
-5.0°C to +105.0°C | |
500 hr. |
±0.5°C | |
For Education | |
-2 to 16 | |
1 or 2 with 2 sets of memorized buffers | |
0.1°C | |
Yes | |
12 VDC | |
Automatic from –5.0 to 105.0°C | |
9V | |
203 mm |