missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEND3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA553659
This item is not returnable.
View return policy
Description
Immunogen sequence: CLPQLNDFFS RFWAQREMED SQPSGQVASF FEAEQVDPGH FLDNKDQEEA LSLDRSSTIA SDHVVDTQDL TEFLDEASSP GEFAVFLLHR LFPELFDHRK LGEQYSCYGD GGKQELDPQR LQIIRNYTEI YFPD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Transcriptional repressor which associates with the NoRC (nucleolar remodeling complex) complex and plays a key role in repressing rDNA transcription. The sumoylated form modulates the stability of the NoRC complex component BAZ2A/TIP5 by controlling its USP21-mediated deubiquitination (PubMed:21914818, PubMed:26100909). Binds to unmethylated major satellite DNA and is involved in the recruitment of the Polycomb repressive complex 2 (PRC2) to major satellites. Stimulates the ERCC6L translocase and ATPase activities (PubMed:28977671). [UniProt]Specifications
BEND3 | |
Polyclonal | |
Unconjugated | |
BEND3 | |
BEN domain containing 3; BEN domain-containing protein 3; Bend3; KIAA1553; mKIAA1553 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57673 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T5X7 | |
BEND3 | |
Recombinant protein corresponding to Human BEND3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only