Learn More
Lenz Laborglasinstrumente™ Bent Adapter
Allows for tubing connections. Lenz Laborglasinstrumente™ Bent Adapters feature a bent glass olive at 90°.
Brand:  Lenz Laborglasinstrumente™ 05285104
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10740232
  • 626.46 SEK / Each
Estimated Shipment: 02-10-2024
to see stock.
Description
Description
  • Constructed from DURAN™ tubing
  • With NS socket
  • Bore: 2.5 mm
Specifications
Specifications
Bent Socket Adapter
Duran Glass
NS 29/32 Upper Socket, 29/32 NS Lower Socket