Learn More
Corning™ Bent Discharge Tubes
Corning™ Bent Discharge Tubes for quick and precise dispensing of acids, bases and solvents
Brand: Corning™ 6880
This item is not returnable.
View return policy
Description
- Bent
- Includes protective sleeve
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Bent | |
FEP, Polypropylene | |
Quick and precise dispensing of acids, bases and solvents | |
Protective Sleeve |
Discharge Tube | |
25 to 100 mL | |
25 to 100 mL Bottle Top Dispenser | |
120 mm |