missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzhydryl isothiocyanate, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
Specifications
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Linear Formula | C6H5CH(NCS)C6H5 |
Quantity | 25 g |
Formula Weight | 225.31 |
Percent Purity | 90% |
Chemical Name or Material | Benzhydryl isothiocyanate |
Safety and Handling
Product Identifier
- Benzhydryl isothiocyanate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'