Learn More
Benzo[b]furan-2-carboxylic acid, 98+%, Thermo Scientific Chemicals
Product Code. 11473057
Click to view available options
Quantity:
1 g
5 g
25 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 496-41-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00005848
InChI Key OFFSPAZVIVZPHU-UHFFFAOYSA-N
Synonym benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid
PubChem CID 10331
IUPAC Name 1-benzofuran-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2O1
Specifications
Melting Point 192°C to 196°C
Boiling Point 310°C to 315°C
Quantity 5 g
Beilstein 124204
Merck Index 14,2561
Formula Weight 162.15
Percent Purity ≥98%
Chemical Name or Material Benzo[b]furan-2-carboxylic acid