missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoic acid, free acid, 99%, MP Biomedicals™
Click to view available options
Quantity:
1 kg
Specifications
CAS | 65-85-0 |
Color | White |
Assay Percent Range | 0.99 |
Molecular Formula | C7H6O2 |
Synonym | benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv |
InChI Key | WPYMKLBDIGXBTP-UHFFFAOYSA-N |
SMILES | C1=CC=C(C=C1)C(=O)O |
IUPAC Name | benzoic acid |
Molecular Weight (g/mol) | 122.123 |
PubChem CID | 243 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'