missing translation for 'onlineSavingsMsg'
Learn More

Benzoin isopropyl ether, 98+%, Thermo Scientific Chemicals

Product Code. 11405277
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6652-28-4
Molecular Formula C17H18O2
Molecular Weight (g/mol) 254.33
MDL Number MFCD00015025
InChI Key MSAHTMIQULFMRG-UHFFFAOYNA-N
Synonym benzoin isopropyl ether, 2-isopropoxy-1,2-diphenylethanone, ethanone, 2-1-methylethoxy-1,2-diphenyl, 2-isopropoxy-2-phenylacetophenone, benzoinisopropylether, acetophenone, 2-isopropoxy-2-phenyl, cbmicro_021426, 2-1-methylethoxy-1,2-diphenylethan-1-one, cambridge id 5476007
PubChem CID 110912
IUPAC Name 1,2-diphenyl-2-propan-2-yloxyethanone
SMILES CC(C)OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 76°C to 79°C
Boiling Point 180°C (10 mmHg)
Quantity 500 g
Beilstein 1977110
Formula Weight 254.33
Percent Purity ≥98%
Chemical Name or Material Benzoin isopropyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.