missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals beta-Casomorphin, human, 95+% Product Code.: 15424679

Thermo Scientific Chemicals beta-Casomorphin, human, 95+%

Product Code. 15424679
25mg
Click to view available options
Quantity:
5mg
25mg
This item is not returnable. View return policy

Product Code. 15424679

Brand: Thermo Scientific Chemicals J62271.MB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An opioid peptide

An opioid peptide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102029-74-3

Specifications

Purity ≥95%
Color White
Content And Storage Store at -20°C
Format Powder
Molecular Weight (g/mol) 864.01
Product Type beta-Casomorphin
Quantity 25mg

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY10J033 Certificate TypeCertificate of Analysis Date28/12/2023 Catalog NumberJ62271.MB,J62271.#0
Lot NumberG14X102 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ62271.MB,J62271.#0
Lot NumberU04H014 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ62271.MB,J62271.#0
Lot NumberI11Z023 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ62271.MB,J62271.#0
Lot NumberM10F027 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ62271.MB,J62271.#0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals beta-Casomorphin, human, 95+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.