Learn More
beta-Defensin 1 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
2105.00 SEK - 18720.00 SEK
Specifications
Antigen | beta-Defensin 1 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17864903
|
Invitrogen
500-P253-50UG |
50μg |
2105.00 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17804963
|
Invitrogen
500-P253-100UG |
100 μg |
2700.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17825533
|
Invitrogen
500-P253-1MG |
1 mg |
18720.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: GNFLTGLGHR SDHYNCVSSG GQCLYSACPI FTKIQGTCYR GKAKCCK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BD-1. Anti-Human BD-1-specific antibody was purified by affinity chromatography employing an immobilized Human BD-1 matrix. Sandwich ELISA: To detect Human BD-1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BD-1 (500-P253Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BD-1. Western Blot: To detect Human BD-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BD-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 1, an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. This gene maps in close proximity to defensin family member, defensin, alpha 1 and has been implicated in the pathogenesis of cystic fibrosis.Specifications
beta-Defensin 1 | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P60022 | |
1672 | |
E.coli-derived Recombinant Human BD-1 (47 a.a.) | |
Antigen affinity chromatography | |
DEFB1 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
AW260221; BD1; BD-1; beta defensin 1; beta-defensin 1; beta-defensin-1; DEFB1; DEFB-1; DEFB101; defensin beta 1; defensin, beta 1; HBD1; rBD-1 | |
DEFB1 | |
Primary | |
-20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.