missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Nicotinamide Adenine Dinucleotide Reduced Disodium Salt MP Biomedicals
Click to view available options
Quantity:
5 g
Specifications
Specifications
CAS | 606-68-8 |
Molecular Formula | C21H27N7Na2O14P2 |
MDL Number | MFCD00036200 |
Quantity | 5 g |
Synonym | beta-nadh disodium salt |
InChI Key | QRGNQKGQENGQSE-WUEGHLCSSA-L |
SMILES | [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
IUPAC Name | disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
Molecular Weight (g/mol) | 709.41 |
PubChem CID | 131673989 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction