missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Sitosterol From Soybeans Practical MP Biomedicals
Shop All MP Biomedical Products
Click to view available options
Quantity:
10 g
Specifications
Specifications
CAS | 83-46-5 |
Molecular Formula | C29H50O |
Quantity | 10 g |
Synonym | beta-sitosterol, sitosterol, cupreol, quebrachol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol |
InChI Key | KZJWDPNRJALLNS-VJSFXXLFSA-N |
SMILES | CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C |
IUPAC Name | (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol |
Molecular Weight (g/mol) | 414.718 |
PubChem CID | 222284 |
ChEBI | CHEBI:27693 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Beta-Sitosterol From Soybeans Practical MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction