missing translation for 'onlineSavingsMsg'
Learn More

Betaine monohydrate, 99+%, for analysis, Thermo Scientific Chemicals

524.24 SEK - 1668.56 SEK

Chemical Identifiers

CAS 590-47-6, 107-43-7
Molecular Formula C5H13NO3
Molecular Weight (g/mol) 135.16
MDL Number MFCD00150010
InChI Key NJZRLXNBGZBREL-UHFFFAOYSA-N
Synonym betaine monohydrate, 2-trimethylammonio acetate hydrate, glycine betaine hydrate, unii-tpu0itr2br, carboxymethyl trimethylammonium hydroxide, tpu0itr2br, bet hydrate, betaine, hydrate, dsstox_cid_29381, dsstox_rid_83496
PubChem CID 134128033
IUPAC Name carboxymethyl(trimethyl)azanium;hydroxide;hydrate
SMILES [OH-].C[N+](C)(C)CC(O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10299400
View Documents
Thermo Scientific Acros
402191000
100 g Glass bottle
524.24 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
10144613
View Documents Promotion Details
Thermo Scientific
402195000
500 g Glass bottle
1668.56 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

590-47-6, 107-43-7
135.16
NJZRLXNBGZBREL-UHFFFAOYSA-N
134128033
[OH-].C[N+](C)(C)CC(O)=O
C5H13NO3
MFCD00150010
betaine monohydrate, 2-trimethylammonio acetate hydrate, glycine betaine hydrate, unii-tpu0itr2br, carboxymethyl trimethylammonium hydroxide, tpu0itr2br, bet hydrate, betaine, hydrate, dsstox_cid_29381, dsstox_rid_83496
carboxymethyl(trimethyl)azanium;hydroxide;hydrate

Specifications

590-47-6, 107-43-7
100.0
11% to 13.5% (140°C, 3 hrs)
99+%
(CH3)3NCH2CO2H·OH
04, 346
betaine monohydrate, 2-trimethylammonio acetate hydrate, glycine betaine hydrate, unii-tpu0itr2br, carboxymethyl trimethylammonium hydroxide, tpu0itr2br, bet hydrate, betaine, hydrate, dsstox_cid_29381, dsstox_rid_83496
[OH-].C[N+](C)(C)CC(O)=O
95% min. (c=10, 430nm) 1cm cell
134128033
99.0 to 101.0% (on dry substance)
Crystalline Powder or Crystals
99.0
White
Authentic
C5H13NO3
MFCD00150010
15, 1181
NJZRLXNBGZBREL-UHFFFAOYSA-N
carboxymethyl(trimethyl)azanium;hydroxide;hydrate
135.16
135.16
Analysis
βine monohydrate
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Betaine monohydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only