Learn More
BIBN 4096
Potent and selective CGRP antagonist
Brand:  Tocris Bioscience 4561/10
Product Code. 18085892
  • 4565.00 SEK / 10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Potent and selective CGRP receptor antagonist (IC50 values are 0.03 and 6.4 nM for human and rat CGRP receptors respectively). Displays high affinity for human CGRP receptors (Ki = 14.4 pM); exhibits no significant affinity for 75 other receptors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
>96%
Store at -20°C
BIBN 4096
204697-65-4
869.66
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BIBN 4096 > 10mg