missing translation for 'onlineSavingsMsg'
Learn More

Biliverdin Dihydrochloride, approx. 80%, MP Biomedicals™

Product Code. 11456691
Click to view available options
Quantity:
25 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 55482-27-4
Molecular Formula C33H36Cl2N4O6
Molecular Weight (g/mol) 655.573
InChI Key GNLHFBAZSKVVCQ-VTURHDOZSA-N
Synonym biliverdin hydrochloride
PubChem CID 132991794
IUPAC Name 3-[(2Z)-2-[[3-(2-carboxyethyl)-5-[(E)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-2-yl]methylidene]-5-[(4-ethenyl-3-methyl-5-oxopyrrol-2-yl)methylidene]-4-methylpyrrol-3-yl]propanoic acid;dihydrochloride
SMILES CC1=C(C(=CC2=C(C(=C(N2)C=C3C(=C(C(=O)N3)C)C=C)C)CCC(=O)O)NC1=CC4=NC(=O)C(=C4C)C=C)CCC(=O)O.Cl.Cl

Specifications

Color Green
Assay Percent Range Approx. 80%
Quantity 25 mg
Formula Weight 655.6
Physical Form Solid
Chemical Name or Material Biliverdin Dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.