missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Biotin Control Diet, Pellet

Biotin Control Diet. Contains egg white spray, corn starch, and salt mixture and vitamin fortification.

9775.00 SEK - 36830.00 SEK

Specifications

Product Type Animal Diet
Autoclavable Autoclavable
Disposable Single-use
Format Pellet
Sterility Non-sterile
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11451472
View Documents
MP Biomedicals
0296041010
10 kg
9775.00 SEK
10kg
Please to purchase this item. Need a web account? Register with us today!
11461472
View Documents
MP Biomedicals
0296041020
20 kg
16930.00 SEK
20kg
Please to purchase this item. Need a web account? Register with us today!
11471472
View Documents
MP Biomedicals
0296041050
50 kg
36830.00 SEK
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Biotin Control Diet
  • Contains egg white spray, corn starch, and salt mixture and vitamin fortification
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Autoclavable
Pellet
Refrigerated
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.