Learn More
Biotin, MedChemExpress
MedChemExpress Biotin (Vitamin B7) is a water-soluble B vitamin and serves as a coenzyme for five carboxylases in humans, involved in the synthesis of fatty acids, isoleucine, and valine, and in gluconeogenesis. Biotin is necessary for cell growth, the production of fatty acids, and the metabolism of fats and amino acids.
Brand: Medchem Express HY-B0511
Description
Biotin (Vitamin B7) is a water-soluble B vitamin and serves as a coenzyme for five carboxylases in humans, involved in the synthesis of fatty acids, isoleucine, and valine, and in gluconeogenesis. Biotin is necessary for cell growth, the production of fatty acids, and the metabolism of fats and amino acids.Specifications
Biotin | |
White | |
C10H16N2O3S | |
Vitamin B7 Vitamin H D-Biotin | |
O=C(O)CCCC[C@@H]1SC[C@]([C@]1([H])N2)([H])NC2=O | |
244.31 | |
Research | |
Metabolism-sugar/lipid metabolism |
58-85-5 | |
Research | |
1 g | |
DMSO : 100 mg/mL (409.32 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.09 mM; Need ultrasonic) | |
244.31 | |
98.0% | |
Powder |
Product Suggestions
Customers who viewed this item also viewed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only