Learn More
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Specifications
Quantity | 500 mg |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in chloroform |
IUPAC Name | (1Z,5Z)-cycloocta-1,5-diene;rhodium;trifluoromethanesulfonate |
Formula Weight | 468.34 |
Physical Form | Crystalline |
Chemical Name or Material | Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X20I011 | Certificate of Analysis | 28/10/2022 | 039289.04,039289.ME |
P15E047 | Certificate of Analysis | 12/11/2021 | 039289.04,039289.ME |
B24Q27 | Certificate of Analysis | 12/11/2021 | 039289.04,039289.ME |
A04L09 | Certificate of Analysis | 12/11/2021 | 039289.04,039289.ME |
D28K28 | Certificate of Analysis | 12/11/2021 | 039289.04,039289.ME |
Safety and Handling
Product Identifier
- Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
RUO – Research Use Only
Product Content Correction
Product Title
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction