Learn More
Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%, Thermo Scientific Chemicals
312.00 SEK - 1206.00 SEK
Chemical Identifiers
CAS | 92466-70-1 |
---|---|
Molecular Formula | C4H4F6O3P+ |
Molecular Weight (g/mol) | 245.037 |
MDL Number | MFCD00063314 |
InChI Key | IMDCVAFSSZPRRM-UHFFFAOYSA-N |
Synonym | bis 2,2,2-trifluoroethyl phosphonate, phosphonic acid bis 2,2,2-trifluoroethyl ester, oxo-bis 2,2,2-trifluoroethoxy phosphanium, phosphonic acid, bis 2,2,2-trifluoroethyl ester, bis trifluoroethyl phosphonate, bis 2,2,2-trifluoroethyl phosphite, bis 2,2,2-trifluoroethyl hydrogen phosphite, oxobis 2,2,2-trifluoroethoxy-??-phosphanylium, bis 2,2,2-trifluoroethyl phosphite, technical grade |
PubChem CID | 6386648 |
IUPAC Name | oxo-bis(2,2,2-trifluoroethoxy)phosphanium |
SMILES | C(C(F)(F)F)O[P+](=O)OCC(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15491858
|
Thermo Scientific Alfa Aesar
H55562.03 |
1 g |
312.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15401868
|
Thermo Scientific Alfa Aesar
H55562.06 |
5 g |
930.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15411868
|
Thermo Scientific Alfa Aesar
H55562.14 |
25 g | N/A | ||||||
Description
Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.
Notes
Store at room temperature. Avoid strong oxidizing agents.
Chemical Identifiers
92466-70-1 | |
245.037 | |
IMDCVAFSSZPRRM-UHFFFAOYSA-N | |
6386648 | |
C(C(F)(F)F)O[P+](=O)OCC(F)(F)F |
C4H4F6O3P+ | |
MFCD00063314 | |
bis 2,2,2-trifluoroethyl phosphonate, phosphonic acid bis 2,2,2-trifluoroethyl ester, oxo-bis 2,2,2-trifluoroethoxy phosphanium, phosphonic acid, bis 2,2,2-trifluoroethyl ester, bis trifluoroethyl phosphonate, bis 2,2,2-trifluoroethyl phosphite, bis 2,2,2-trifluoroethyl hydrogen phosphite, oxobis 2,2,2-trifluoroethoxy-??-phosphanylium, bis 2,2,2-trifluoroethyl phosphite, technical grade | |
oxo-bis(2,2,2-trifluoroethoxy)phosphanium |
Specifications
92466-70-1 | |
43°C to 44°C (2 mmHg) | |
C4H4F6O3P+ | |
1.332 | |
bis 2,2,2-trifluoroethyl phosphonate, phosphonic acid bis 2,2,2-trifluoroethyl ester, oxo-bis 2,2,2-trifluoroethoxy phosphanium, phosphonic acid, bis 2,2,2-trifluoroethyl ester, bis trifluoroethyl phosphonate, bis 2,2,2-trifluoroethyl phosphite, bis 2,2,2-trifluoroethyl hydrogen phosphite, oxobis 2,2,2-trifluoroethoxy-??-phosphanylium, bis 2,2,2-trifluoroethyl phosphite, technical grade | |
C(C(F)(F)F)O[P+](=O)OCC(F)(F)F | |
245.037 | |
246.05 | |
Technical | |
Bis(2,2,2-trifluoroethyl) phosphite |
1.545 g/mL | |
76°C (169°F) | |
(CF3CH2O)2P(O)H | |
MFCD00063314 | |
IMDCVAFSSZPRRM-UHFFFAOYSA-N | |
oxo-bis(2,2,2-trifluoroethoxy)phosphanium | |
6386648 | |
90% | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.