missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl) ether, 99%, Thermo Scientific Chemicals

Product Code. 11462347
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

Solubility
Soluble in water. (1.1 g/L) at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-44-4
Molecular Formula C4H8Cl2O
Molecular Weight (g/mol) 143.01
MDL Number MFCD00000975
InChI Key ZNSMNVMLTJELDZ-UHFFFAOYSA-N
Synonym 2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether
PubChem CID 8115
ChEBI CHEBI:34573
IUPAC Name 1-chloro-2-(2-chloroethoxy)ethane
SMILES ClCCOCCCl

Specifications

Melting Point -50°C
Density 1.219
Boiling Point 178°C
Flash Point 55°C (131°F)
Odor Pungent
Linear Formula ClCH2CH2OCH2CH2Cl
Refractive Index 1.457
Quantity 1000 g
UN Number UN1916
Beilstein 605317
Merck Index 14,3066
Solubility Information Soluble in water. (1.1g/L) at 20°C.
Formula Weight 143.01
Percent Purity 99%
Chemical Name or Material Bis(2-chloroethyl) ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bis(2-chloroethyl) ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Carcinogenicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : Bis(2-chloroethyl) ether

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'