missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis[2-(diphenylphosphino)ethyl]ammonium chloride, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is involved in the Synthesis of the new chiral aminodiphosphine ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is involved in the Synthesis of the new chiral aminodiphosphine ligands.
Solubility
Insoluble in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
It is involved in the Synthesis of the new chiral aminodiphosphine ligands.
Solubility
Insoluble in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 175°C to 180°C |
Linear Formula | [(C6H5)2PCH2CH2]2NH2Cl |
Quantity | 5 g |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 477.95 |
Percent Purity | ≥97% |
Chemical Name or Material | Bis[2-(diphenylphosphino)ethyl]ammonium chloride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction