Learn More
Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%, Thermo Scientific Chemicals
775.56 SEK - 2531.98 SEK
Chemical Identifiers
CAS | 23582-02-7 |
---|---|
Molecular Formula | C34H33P3 |
Molecular Weight (g/mol) | 534.56 |
MDL Number | MFCD00003048 |
InChI Key | AXVOAMVQOCBPQT-UHFFFAOYSA-N |
Synonym | bis 2-diphenylphosphinoethyl phenylphosphine, phenylphosphinediyl bis ethane-2,1-diyl bis diphenylphosphine, bis 2-diphenylphosphino ethyl phenylphosphine, phosphine, bis 2-diphenylphosphino ethyl phenyl, phenylbis diphenylphosphinoethyl phosphine, pubchem6540, acmc-20alo6, chembl69711, bis 2-biphenylphosphinoethyl phenylphosphine |
PubChem CID | 90192 |
IUPAC Name | bis(2-diphenylphosphanylethyl)-phenylphosphane |
SMILES | C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10003600
|
Thermo Scientific Acros
316820010 |
1 g | Glass bottle |
775.56 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10301284
|
Thermo Scientific Acros
316820050 |
5 g | Glass bottle |
2531.98 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23582-02-7 | |
534.56 | |
AXVOAMVQOCBPQT-UHFFFAOYSA-N | |
90192 | |
C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
C34H33P3 | |
MFCD00003048 | |
bis 2-diphenylphosphinoethyl phenylphosphine, phenylphosphinediyl bis ethane-2,1-diyl bis diphenylphosphine, bis 2-diphenylphosphino ethyl phenylphosphine, phosphine, bis 2-diphenylphosphino ethyl phenyl, phenylbis diphenylphosphinoethyl phosphine, pubchem6540, acmc-20alo6, chembl69711, bis 2-biphenylphosphinoethyl phenylphosphine | |
bis(2-diphenylphosphanylethyl)-phenylphosphane |
Specifications
23582-02-7 | |
100.0 | |
White | |
Authentic | |
C34H33P3 | |
bis 2-diphenylphosphinoethyl phenylphosphine, phenylphosphinediyl bis ethane-2,1-diyl bis diphenylphosphine, bis 2-diphenylphosphino ethyl phenylphosphine, phosphine, bis 2-diphenylphosphino ethyl phenyl, phenylbis diphenylphosphinoethyl phosphine, pubchem6540, acmc-20alo6, chembl69711, bis 2-biphenylphosphinoethyl phenylphosphine | |
AXVOAMVQOCBPQT-UHFFFAOYSA-N | |
bis(2-diphenylphosphanylethyl)-phenylphosphane | |
90192 | |
97% | |
Bis(2-diphenylphosphinoethyl)phenylphosphine |
96.0 | |
121.0°C to 126.0°C | |
74.1 to 78.7% (C) | |
97% | |
MFCD00003048 | |
Solubility in water: insoluble | |
C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 | |
534.56 | |
534.56 | |
Chunks and/or Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 245-753-8
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.