Learn More
Bis(cyclopentadienyl)zirconium chloride hydride, 95%, Thermo Scientific Chemicals
601.15 SEK - 7135.76 SEK
Chemical Identifiers
CAS | 37342-97-5 |
---|---|
Molecular Formula | C10H11ClZr |
Molecular Weight (g/mol) | 257.86 |
MDL Number | MFCD02089401 |
InChI Key | VPQMECJLNPUNGN-UHFFFAOYSA-M |
Synonym | zirconocene chloride hydride |
PubChem CID | 131718033 |
IUPAC Name | cyclopenta-1,3-diene;hydride;zirconium(2+);chloride |
SMILES | [H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10391234
|
Thermo Scientific Acros
207890010 |
1 g | Glass bottle |
601.15 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10236790
|
Thermo Scientific Acros
207890050 |
5 g | Glass bottle |
1763.23 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10762315
|
Thermo Scientific Acros
207890250 |
25 g | Glass bottle |
7135.76 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
37342-97-5 | |
257.86 | |
VPQMECJLNPUNGN-UHFFFAOYSA-M | |
131718033 | |
[H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2] |
C10H11ClZr | |
MFCD02089401 | |
zirconocene chloride hydride | |
cyclopenta-1,3-diene;hydride;zirconium(2+);chloride |
Specifications
37342-97-5 | |
100.0 | |
White to Beige | |
94% min. (Argentometry) | |
MFCD02089401 | |
zirconocene chloride hydride | |
VPQMECJLNPUNGN-UHFFFAOYSA-M | |
cyclopenta-1,3-diene;hydride;zirconium(2+);chloride | |
131718033 | |
95% | |
Bis(cyclopentadienyl)zirconium chloride hydride |
94.0 | |
>300.0°C | |
Authentic | |
C10H11ClZr | |
15, 8533 | |
Solubility in water: reacts with water. | |
[H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2] | |
257.86 | |
257.86 | |
Powder |
Safety and Handling
GHS H Statement
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 253-479-5
- Bis(cyclopentadienyl)zirconium chloride hydride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H261-In contact with water releases flammable gases.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only