missing translation for 'onlineSavingsMsg'
Learn More

Bis(tricyclohexylphosphine)palladium(0), 97% min, Thermo Scientific Chemicals

Air Sensitive

1655.00 SEK - 5640.00 SEK

Chemical Identifiers

CAS 33309-88-5
Molecular Formula C36H66P2Pd
Molecular Weight (g/mol) 667.29
MDL Number MFCD01073796
InChI Key JGBZTJWQMWZVNX-UHFFFAOYSA-N
Synonym bis tricyclohexylphosphine palladium 0, bis tricyclohexylphosphine palladium o, bis tricyclohexylphosphine palladium, palladium, bis tricyclohexylphosphine, pd pcy3 2, palladium; tricyclohexylphosphane, palladium-tricyclohexylphosphine 1:2, bis tricyclohexylphosphine-palladium oShow MoreShow Less
PubChem CID 2734559
IUPAC Name palladium;tricyclohexylphosphane
SMILES [Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15407075
SDS View Documents
Thermo Scientific Alfa Aesar
045792.MD
250 mg
1655.00 SEK
250mg
Please to purchase this item. Need a web account? Register with us today!
15402457
SDS View Documents
Thermo Scientific Alfa Aesar
045792.03
1 g
5640.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bis(tricyclohexylphosphine)palladium(0) is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(tricyclohexylphosphine)palladium(0), 97% min, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.