missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Blue Detectable Volumetric Spoon

Blue detachable spoons feature a special additive in the material that makes them visible to metal detectors or X rays.

Brand:  Buerkle™ 5378-4099

2033.58 SEK valid until 2024-12-31
Use promo code "23267" to get your promotional price.



Additional Details : Weight : 2.10000kg

 View more versions of this product

Product Code. 16515260

  • 2225.00 SEK / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

  • Volumetric and dosing spoons can be used to measure small amounts of powder, granules and liquids exactly and dose them precisely
  • For precise measuring, excess medium is stripped off the upper edge of the spoon
  • The content volume is engraved on the handle
  • The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing for good emptying of small remaining amounts
  • The flat base makes it possible to set the volumetric spoon down without spilling the content
  • The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities
  • Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded
  • The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed and the food and beverage industry
  • Individually packaged
  • Can be detected by color scanners, metal detectors or X-rays
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • With stripper edge
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
Specifications

Specifications

Sampler
Dosing spoons with stripper edge for exact measuring
Yes
Blue
Especially for The Food, Animal Feed and Pharmaceutical Industries
Polystyrene
50 mL
Sterile
Food Contact, Reach, RoHS
100
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers