Learn More
Brady™ BMP71 BMP61 M611 Permanent Vinyl General Identification Labels
Permanent Vinyl (B-439) is a thermal transfer printable material ideal for applications needing various colors like finished product ID, rating plates and general purpose ID.
Brand:  Brady™ 018620
 View more versions of this product
Product Code. 11549033
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Labels for the BMP61 and M611 Thermal Transfer Portable Printers
  • Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Specifications
Permanent Acrylic
Gloss
Thermal Transfer
15.24 m x 48.26 mm
White
Polyester (B-439)
BMP61, BMP71 (with Media Adapter), M611