Learn More
BMS-754807, MedChemExpress
MedChemExpress BMS-754807 is a potent and reversible IGF-1R/IR inhibitor (IC50=1.8 and 1.7 nM, respectively; Ki=IC50 values of 6, 44, 7, 4, 9, and 25 nM, respectively.
Brand: Medchem Express HY-10200
This item is not returnable.
View return policy
Description
BMS-754807 is a potent and reversible IGF-1R/IR inhibitor (IC50=1.8 and 1.7 nM, respectivelySpecifications
BMS-754807 | |
Off-White | |
C23H24FN9O | |
DMSO : ≥ 100 mg/mL (216.69 mM) | |
461.49 | |
98.74% | |
Solid |
1001350-96-4 | |
Research | |
10 mg | |
FC1=CC=C(NC([C@@]2(C)N(C(N=C3NC4=NNC(C5CC5)=C4)=NN6C3=CC=C6)CCC2)=O)C=N1 | |
461.49 | |
Research | |
Cancer-Kinase/protease |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.