Learn More
Bohlender™ BOLA™ Conductors for Magnetic Stirrer Heads
Conductors for Magnetic Stirrer Heads made of borosilicate glass.
Brand: Bohlender™ C463-19
This item is not returnable.
View return policy
Description
- Borosilicate glass with ground joint and glued upper bearing
- Alternatively without ground joint for fusing on the reactor lid (upper bearing to be glued is included).
Specifications
NS 19/26 | |
NS 19/26 | |
Borosilicate Glass | |
Left-handed Thread M 14.5 x 1.5 |