Learn More
Bohlender™ BOLA™ Laboratory Screw Joints EX
Laboratory screw joints EX made of conductive PPS-EX with glass fiber and PTFE-EX.
Brand: Bohlender™ D842-62
This item is not returnable.
View return policy
Description
- Screw cap made of conductive black PPS-EX reinforced with glass fibers
- Inner parts made of conductive PTFE-EX
Specifications
Screw Joint | |
0.31 in. | |
GL 25 |
PPS-EX, PTFE-EX | |
8 mm | |
Black |