Learn More
Bohlender™ BOLA™ Laboratory Screw Joints EX
Laboratory screw joints EX made of conductive PPS-EX with glass fiber and PTFE-EX.
Brand:  Bohlender™ D842-62
 View more versions of this product
Product Code. 10360045
  • 701.00 SEK / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Screw cap made of conductive black PPS-EX reinforced with glass fibers
  • Inner parts made of conductive PTFE-EX
Specifications
Specifications
Screw Joint
0.31 in.
GL 25
PPS-EX, PTFE-EX
8 mm
Black