Learn More
Bohlender™ BOLA™ Laboratory Screw Joints EX
Laboratory screw joints EX made of conductive PPS-EX with glass fiber and PTFE-EX.
Brand:  Bohlender™ D841-74
 View more versions of this product
Product Code. 10594555
  • 419.76 SEK / Each
Estimated Shipment: 11-11-2024
to see stock.
Explore more special offers
Description
  • Screw cap made of conductive black PPS-EX reinforced with glass fibers
  • Inner parts made of conductive PTFE-EX
Specifications
Specifications
Screw Joint
0.39 in.
GL 18
PPS-EX, PTFE-EX
10 mm
Black