Learn More
Bohlender™ Bola PTFE/PEEK Propeller with 4 Blades
Brand:  Bohlender™ C448-08
 View more versions of this product
Product Code. 10832100
  • 1315.00 SEK / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Specifications
8 mm
4 Blades
-200°C to +250°C
50 L x 22 W x 4 Hmm
The product is sucked bottom-up, good axial flow with low shear force.
1 Each
Propeller