Learn More
Robu™ Borosilicate Glass Buchner Filter Funnels
1080.00 SEK - 1085.00 SEK
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | Highly resistant to most chemicals such as solvents, acids and alkalis |
Product Code | Brand | Capacity (Metric) | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Pore Size | Price | Quantity & Availability | ||||
10614472
|
Robu™
21552 |
500 mL | 2 |
1080.00 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
10151020
|
Robu™
21253 |
250 mL | 3 |
1085.00 SEK
Each |
In Stock
Log in to see stock. |
||||
Specifications
Borosilicate Glass | |
Highly resistant to most chemicals such as solvents, acids and alkalis |