Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Removable Fritted Disc
6952.00 SEK - 7900.00 SEK
Specifications
Diameter (Metric) Inner | 20 mm |
---|---|
Type | Chromatography Column |
For Use With (Application) | Ion exchange chromatography |
Quantity | 1 Each |
USP Type | Type I |
Product Code | Brand | Column Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Capacity | Price | Quantity & Availability | |||||
11890522
|
Kimble Chase™
422230-2520 |
75 mL |
7900.00 SEK
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
11880522
|
Kimble Chase™
422230-5020 |
150 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
20 mm | |
Ion exchange chromatography | |
Type I | |
With removable fritted disc |
Chromatography Column | |
1 Each | |
Borosilicate Glass |