Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Filter Funnels
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Filter Funnel is a laboratory funnel used for separating solids from liquids via the process of filtration. In order to achieve this, a cone-like shaped piece of filter paper is usually folded into a cone and placed within the funnel.
Brand:  Lenz Laborglasinstrumente™ 07200425
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10548421
  • 394.32 SEK / Each
Estimated Shipment: 31-07-2024
to see stock.
Add to basket
Description
Description
  • Conical shaped filter funnel
  • Made from borosilicate glass
Specifications
Specifications
25 mL
100 mm
4
Borosilicate Glass
25mm
Filter Funnel