Learn More
Fisherbrand™ Borosilicate Glass Conical Shape Narrow Neck Erlenmeyer Flasks
Brand: Fisherbrand™ 66593480
This item is not returnable.
View return policy
Description
Erlenmeyer, graduated.Specifications
ISO 1773 | |
5 L | |
Conical |
50 mm | |
Borosilicate Glass | |
Narrow |