Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 01400045
Additional Details : Weight : 1.00000kg
Description
- Acc. to DIN 12252, form C
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Hollow Stopper | |
5/Pk. | |
Borosilicate Glass | |
NS 45/40 |