Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01400007
Additional Details : Weight : 0.00200kg
 View more versions of this product
Product Code. 10291631
  • 244.86 SEK / Pack of 10
Estimated Shipment: 31-07-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12252, form C
  • Available in a range of cone sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Hollow Stopper
10/Pk.
Borosilicate Glass
NS 7/16