Learn More
Kimble Chase™ Borosilicate Glass Serum Vials without Closures
Kimble Chase™ Borosilicate Glass Serum Vials without Closures are ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, vaccines, blood plasma, culture media, chromatography samples, and more.
Brand: Kimble Chase™ 62113U-2
Additional Details : Weight : 10.00000kg
Description
- Special design provides extra strength for freeze-drying applications
- Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
- Special blowback design provides ease of use for lyophillization stoppers
- Autoclavable
- Aluminum seals, stoppers and septa are available separately
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
15 mm | |
2 mL | |
32 mm |
Borosilicate Glass (Type 1 Class A) | |
4640 | |
A/S 13 |