Learn More
Borosilicate Glass Stopcock Adapter with Cone
Control flow rate. Quickfit Jointed Ware™ QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
Brand:  Quickfit MF11/3/SC
 View more versions of this product
Product Code. 11322045
  • 964.00 SEK / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Description
Control flow rate. The QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
  • With GP Rotaflo™ PTFE stopcock: - No lubrication required - No contaminating greases - Fine flow control - Positive closure
  • Detachable plastic screw thread connectors allow rubber hose etc. , to be fitted easily and safely
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
24/29
Cone, screw thread adapter and GP Rotaflo™ PTFE stopcock
Stopcock Adapter