missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ Bottle Set for Titration Agent
SI Analytics™ Bottle Set for Titration Agent offers a safe and secure place to store titration agents.
Brand: SI Analytics™ 285221047
This item is not returnable.
View return policy
Description
- Holds up to 1L
- Square brown bottle
- Includes bottle attachment with GL 45 thread
Specifications
Brown | |
Reagent bottle | |
1 L |
Bottle Top Adapter | |
GL45 |