Learn More
Trajan Scientific and Medical™ BP624 GC Capillary Columns
Used for EPA volatile organics analysis. Trajan Scientific and Medical BP624 GC Capillary Columns are polar phase designed columns.
Brand:  Trajan Scientific and Medical™ 054842
 View more versions of this product
Product Code. 15278713
  • 14695.00 SEK / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Polar phase
  • EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
  • Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
Specifications
0.25 mm
Drinking Water Volatiles Analysis, Drinking Water Volatiles, Chlorinated Hydrocarbons, Solvents Analysis
1.4 μm
Cyanopropylphenyl Polysiloxane
BP624 GC Capillary Column
0°C to 230/240°C
60 m
BP624