Learn More
Brady™ FreezerBondz II Polyester Laboratory Labels B490
FreezerBondz II Matte White Polyester Laboratory Labels are durable polyester markers designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand: Brady™ 143089
1558.00 SEK valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
- White polyester material has a permanent acrylic adhesive and is thermal transfer printable
- Good print smudge resistance, solvent resistance and high/low temperature performance of -320 to 212° F (-196 to 100° C)
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
- Brady Series R4300 ribbon is recommended
Specifications
RoHS, 2002/618/EC MCV, 2002895/EC | |
White | |
Laboratory Label | |
For use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides | |
1500/Pk. | |
0.6 x 1.625 in. | |
41.28 mm | |
15.20 mm |
Acrylic, Permanent | |
Polyester | |
Non-sterile | |
BBP™33 Printer | |
0.0021 in. | |
1.6 in. | |
0.6 in. |