Learn More
Brady™ StainerBondz™ Tissue Cassette Labels
Give histology and anatomical pathology labs a reliable solution for lab labeling and specimen tracking
Brand:  Brady™ 804865
 View more versions of this product
Product Code. 11588844
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Tissue cassette labels can withstand tissue processing protocols and up to 20 years in storage
  • Durability and print quality that histology and anatomic pathology labs need for accurate, identifiable specimen labeling
  • Labels must be used with the BSP 31 Label Attachment System
Specifications
Specifications
RoHS
Acrylic, Permanent
Matte
Polyester
General identification, Laboratory identification
1
Rectangle
1 in.
0.25 in.
IDXPERT Tissue Cassette Labels
White
Label
StainerBondz™ Tissue Cassette Label
LabXpert™ Printer
450/Pk.
0.117 mm
25.50 mm
0.6 cm