Learn More
Brady™ BradyGrip™ Print-on-Hook Labels featuring VELCRO™ Hook for M7 Printers
For wire and cable ID, BradyGrip™'s Print-on Hook Material is a unique print-and-place material that can be easily applied, repositioned or removed.
Brand:  Brady™ 173230
 View more versions of this product
Product Code. 17819044
  • 362.00 SEK / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Features a printable front and grippy back with tiny hooks
  • Attaches to VELCRO™ Brand ONE-WRAP™ Strap for easy wire and cable ID
  • Doesn't damage wires and cables like nylon cable ties
  • Maximizes your uptime with faster, printable ID on cable bundles
  • VELCRO™ Brand ONE-WRAP™ product not included with purchase
Specifications
Specifications
Not Autoclavable
None
Matte
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
20 ft.
-40°F to +122°F
0.015 in.
20 ft. x 0.5 in.
6.1 m
12.7 mm
Non-adhesive
White
Continuous
Single-side Printable
Labels
Wire and Cable Labeling
No
Rectangle
-40°C to +50°C
0.381 mm
20 ft.
0.5 in.