Learn More
Brady™ BradyJet J2000 Color Label Printer Kit EU with Workstation SFID Suite
The BradyJet J2000 Color Label Printer Kit allows you to print in full color and high resolution with the included Workstation SFID Suite.
Brand: Brady™ 199968
21321.56 SEK valid until 2024-12-31
Use promo code "21918" to get your promotional price.
Additional Details : Weight : 3.30000kg
Description
The BradyJet J2000 Color Label Printer Kit EU with Brady™ Workstation SFID Suite works great in a multitude of industries, from lab to safety and more.
- Compact design means it can fit in tight spaces
- On-demand, mid-volume inkjet solution creates photo-quality color labels in a single pass
- More color options than you'd ever need- up to 16 million
- Custom Identification without the high cost of outsourced printing
- Setup is stress free- Contact our Brady™ Technical Support Team to schedule a free phone appointment
Specifications
Inkjet | |
264 mm | |
180 mm | |
389 mm | |
Non-sterile | |
J2000 Printer, EU Power Cord with AC Adapter, USB Cable, Quick Start Guide, Ink Cartridge, Material Roll for Printhead Alignment, USB Stick with Drivers and Manuals, Brady™ Workstation SFID Suite | |
J2000 | |
4800 dpi | |
Windows™ 7, Windows™ 8, Windows™ 10 | |
7 lb. | |
Barcode and Electrical Labeling, Hazard Identification, Laboratory Labeling | |
110/240 V |
10.4 in. | |
7 in. | |
15.3 in. | |
Not Autoclavable | |
Multi-color | |
Chinese (Tradditional), Chinese (Simplified), English, French, German, Italian, Japanese, Korean, Spanish | |
63.5 mm/s | |
USB | |
15°C to 32°C | |
3.2 kg | |
1 Year |