Learn More
BRAND™ BRANDplates™ PC Membrane Single Inserts
Sterile, individual inserts
Brand: BRAND™ 782866
3456.19 SEK valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.07600kg
Description
Specifications
Smooth walled | |
24-/6-well plates BRANDplates™ | |
48 |
PC Membrane 8.0 μm | |
13 mm |