Learn More
BrandTech™ accu-jet™ pro Pipet Controller
Improves upon the original accu-jet with new features and functionality
POA
Specifications
Autoclavable | Not Autoclavable |
---|---|
Battery Type | NiMH |
Type | Pipette Controller |
Electrical Requirements | 120V, 60Hz |
Grip Type | Ergonomic |
Description
Range dial allows you to set maximum pipetting speed, but still provides the touch-sensitive, continuously adjustable speed control of the original accu-jet.
- Switch-selectable gravity or powered dispensing offers outstanding volumetric pipetting
- Soft blow-out control minimizes disturbance of cell pellets, or density gradient interfaces
- Reshaped, ergonomic grip
- NiHM battery pack, wall holder, 120V 60Hz battery charger, and two extra 0.2μm hydrophobic filters are included
Warranty
One year
Specifications
Not Autoclavable | |
Pipette Controller | |
Ergonomic | |
NiMH battery pack, wall holder, 120V 60Hz battery charger and two extra 0.2μm hydrophobic filters | |
50°F to 104°F | |
120 V |
NiMH | |
120V, 60Hz | |
60 Hz | |
0.2μm | |
10°C to 40°C | |
1 Year |