missing translation for 'onlineSavingsMsg'
Learn More

BRCA2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

1422.00 SEK - 4180.00 SEK

Specifications

Antigen BRCA2
Concentration 0.5 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17783584
View Documents
Invitrogen
PA5122190
20 μL
1422.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
17773584
View Documents
Invitrogen
PA5122189
100 μL
4180.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Breast Cancer Type 2 susceptibility protein (BRCA2) is a tumor-suppressor protein reported to be involved in double-strand break repair, homologous recombination and S-phase checkpoint activation.
Specifications

Specifications

BRCA2
Western Blot
Unconjugated
Rabbit
Human
P51587
675
Lys2308-His2537 (Accession P51587), with N-terminal His Tag
Primary
-20°C
BRCA2
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300
brca 2; BRCA1/BRCA2-containing complex, subunit 2; BRCA2; brca-2; BRCA2 DNA repair associated; BRCA2, DNA repair associated; BRCC2; breast and ovarian cancer susceptibility gene, early onset; breast and ovarian cancer susceptibility protein 2; breast cancer 2; breast cancer 2 tumor suppressor; breast cancer 2, early onset; breast cancer 2, mutation 1, University of Wisconsin-Madison; breast cancer susceptibility protein 2; breast cancer type 2 susceptibility protein; Breast cancer type 2 susceptibility protein homolog; BROVCA2; FACD; FAD; FAD1; FANCB; FANCD; FANCD1; Fanconi anemia group D1 protein; Fanconi anemia group D1 protein homolog; GLM3; mutant BRCA2; PNCA2; RAB163; RP11-298P3.4; XRCC11
BRCA2
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications