missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ BrdU Staining Kit for Flow Cytometry FITC

Contains the necessary reagents and buffers for identifying and examining proliferating cells of mammalian species by flow cytometric analysis. This kit is optimized to achieve brighter staining compared to traditional protocols.

Brand:  Invitrogen™ 8811-6600-42

Product Code. 15581287

  • 7770.00 SEK / 100 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

Contains Sodium azide and formaldehyde


Includes:
  • BrdU
  • DNase I
  • Anti-BrdU FITC Antibody (clone BU20A)
  • BrdU Staining Buffer Concentrate (4X)
  • Fixation/Permeabilization Diluent
This item is not returnable. View return policy

Description

Description

  • Cycling cells are incubated with 5-bromo-2′-deoxyuridine (BrdU), a synthetic analog of thymidine which incorporates into newly synthesized genomic DNA during the S-phase of mitosis
  • Following DNA denaturation, the cells are stained for BrdU incorporation along with any other cell surface and/or intracellular targets of interest
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fluorescence
Flow Cytometer
Kit
100 Tests
BrdU Staining
Staining Kit
eBioscience™
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ eBioscience™ BrdU Staining Kit for Flow Cytometry FITC > BrdU Staining Kit for Flow Cytometry FITC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.