missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BSX Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1925.00 SEK - 4460.00 SEK
Specifications
Antigen | BSX |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18398884
|
Bio-Techne
NBP3-10602-25UL |
25 μg |
1925.00 SEK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18321604
|
Bio-Techne
NBP3-10602-100UL |
100 μg |
4460.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BSX Polyclonal specifically detects BSX in Mouse samples. It is validated for Western Blot.Specifications
BSX | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
brain-specific homeobox, brain-specific homeobox protein homolog, BSX1 | |
The immunogen is a synthetic peptide directed towards the middle region of MOUSE BSX (NP_839976). Peptide sequence RRMKHKKQLRKSQDEPKAADGPESPEGSPRAPEGAPADARLSLPAGAFVL | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
390259 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title